missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calcium Acetate, Monohydrate, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

Manufacturer:  Avantor Performance 126601

 View more versions of this product

Catalog No. 02002509



Specifications

Specifications

Calcium Acetate
5743-26-0
calcium acetate monohydrate, calcium diacetate monohydrate, unii-7za48gim5h, calcium diacetate hydrate, acetic acid, calcium salt, monohydrate, calcium acetate, monohydrate, 7za48gim5h, calcium acetat, calcium acetate jan, acmc-1c5pu
CC(=O)[O-].CC(=O)[O-].O.[Ca+2]
176.181
CHEBI:59199
Powder
Wide Mouth Amber Glass Bottle
500g
Monohydrate
C4H8CaO5
XQKKWWCELHKGKB-UHFFFAOYSA-L
calcium;diacetate;hydrate
82163
176.19
BAKER ANALYZED™ ACS Reagent
1L = 1.50kg

Chemical Identifiers

5743-26-0
176.181
calcium acetate monohydrate, calcium diacetate monohydrate, unii-7za48gim5h, calcium diacetate hydrate, acetic acid, calcium salt, monohydrate, calcium acetate, monohydrate, 7za48gim5h, calcium acetat, calcium acetate jan, acmc-1c5pu
CHEBI:59199
CC(=O)[O-].CC(=O)[O-].O.[Ca+2]
C4H8CaO5
XQKKWWCELHKGKB-UHFFFAOYSA-L
82163
calcium;diacetate;hydrate
Documents