missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Calcium acetate hydrate, 99%, extra pure, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 211052500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Calcium acetate hydrate | |
114460-21-8 | |
Powder and/or Agglomerates | |
98.5 to 101.5% (on anhydrous substance) (Complexometry) | |
(CH3CO2)2Ca | |
02,IV,113 | |
14,1646 | |
Solubility in water: 250g/L (20°C). Other solubilities: slightly soluble in alcohol | |
[Ca++].CC([O-])=O.CC([O-])=O | |
158.17 | |
99% | |
Authentic |
White | |
7.0 to 9.0 (10% soln., 20°C) | |
250 g | |
C4H6CaO4 | |
MFCD00012448 | |
13,60; 15,46 | |
Acetic acid, calcium salt hydrate | |
VSGNNIFQASZAOI-UHFFFAOYSA-L | |
calcium diacetate | |
158.17 | |
Extra Pure | |
Plastic bottle |
Chemical Identifiers
114460-21-8 | |
158.17 | |
VSGNNIFQASZAOI-UHFFFAOYSA-L | |
calcium diacetate |
C4H6CaO4 | |
MFCD00012448 | |
Acetic acid, calcium salt hydrate | |
[Ca++].CC([O-])=O.CC([O-])=O |
RUO – Research Use Only