missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Calcium acetate hydrate, 99%, extra pure, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 211050051
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Calcium acetate hydrate | |
5kg | |
C4H6CaO4·xH2O | |
02,IV,113 | |
14,1646 | |
Solubility in water: 250g/L (20°C). Other solubilities: slightly soluble in alcohol | |
158.17 | |
Extra Pure | |
Plastic drum |
114460-21-8 | |
98.5 to 101.5% (on anhydrous substance) (Complexometry) | |
(CH3CO2)2Ca | |
13,60; 15,46 | |
Acetic acid, calcium salt hydrate | |
158.17 | |
99% | |
Authentic |
RUO â Research Use Only