missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Calcium acetate hydrate, 99%

Manufacturer:  Alfa Aesar™ A1494722

 View more versions of this product

Catalog No. AAA1494722



Description

Description

Applications
Calcium acetate hydrate is used as a precursor to produce calcium ions in the development of calcium oxide based mesoporous carbon dioxide sorbent. It is an active component of pyroligneous acid-modified limestone. It is employed in the preparation of magnetite-calcium phosphate core-shell nanoparticles. It serves as a raw material in the preparation of hydroxyapatite-wollastonite composites. Further, it acts as a crosslinking agent in the microfluidic production of alginate hydrogel microbeads. In addition to this, it is used to induce crystallization of proteins.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

Calcium acetate hydrate
114460-21-8
Ca(OOCCH3)2·xH2O
3692527
158.17 (Anhydrous)
100g
C4H6O4Ca
MFCD00012448
14,1646
99%

Chemical Identifiers

114460-21-8
C4H6O4Ca
MFCD00012448
Safety and Handling

Safety and Handling

RTECSNumber : AF7525000

TSCA : TSCA

Documents