missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calcium, AAS standard solution, Specpure™, Ca 1000μg/mL, Alfa Aesar™

Manufacturer:  Alfa Aesar 88057AP

 View more versions of this product

Catalog No. AA88057AP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Calcium
CCaO3
Nitric acid: 5%; Calcium carbonate: 0.25%; Water: 94.75%
C(=O)([O-])[O-].[Ca+2]
100.086
CHEBI:3311
17mm Hg at 20°C
500mL
471-34-1,7732-18-5,7732-18-5
MFCD00085314
VTYYLEPIZMXCLO-UHFFFAOYSA-L
calcium;carbonate
10112
Liquid
Colorless

Chemical Identifiers

471-34-1
100.086
VTYYLEPIZMXCLO-UHFFFAOYSA-L
CHEBI:3311
C(=O)([O-])[O-].[Ca+2]
CCaO3
MFCD00085314
10112
calcium;carbonate
Safety and Handling

Safety and Handling

H314: Causes severe skin burns and eye damage.

DOTInformation : DOT Class: 8, Packaging Group: III

EINECSNumber : 231-714-2

SDS Documents