Please login to your online account to display your discounted pricing

Calcium, 99%, turnings, ACROS Organics™


Quantity 500g
Packaging Plastic bucket

Manufacturer: thermo fisher scientific  AC365745000

 View more versions of this product

Catalog No. AC365745000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5% (Titrimetry other)
Formula Weight 40.07
Physical Form Liquid
Quantity 500g
Color Clear colorless to pale yellow
Merck Index 15, 1648
Fieser 05,89; 09,1634; 14,71
Total Trace Metal Impurities 1 % max.
Health Hazard 1 GHS Signal Word: Danger
Molecular Formula Ca
Chemical Name or Material Calcium
Assay Percent Range 99%
Linear Formula Ca
Packaging Plastic bucket
Name Note 99%
CAS 7440-70-2
MDL Number MFCD00085314
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Handle under inert gas. Protect from moisture.
Keep away from any possible contact with water, because of violent reaction and possible flash fire.
Brush off loose particles from skin. Immerse in cool water/wrap in wet bandages.
In case of fire: Use CO2, dry chemical, or foam.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
In contact with water releases flammable gases.

Specifications

Chemical Name or Material Calcium
Name Note 99%
CAS 7440-70-2
Physical Form Liquid
Assay ≥98.5% (Titrimetry other)
Assay Percent Range 99%
Molecular Formula Ca
Linear Formula Ca
Formula Weight 40.07
MDL Number MFCD00085314
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Handle under inert gas. Protect from moisture.
Keep away from any possible contact with water, because of violent reaction and possible flash fire.
Brush off loose particles from skin. Immerse in cool water/wrap in wet bandages.
In case of fire: Use CO2, dry chemical, or foam.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
In contact with water releases flammable gases.

SDS & Safety Documents