missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calcium, 99%, turnings, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC365741000

 View more versions of this product

Catalog No. AC365741000


 

Specifications Description & Specifications

Specifications

≥98.5% (Titrimetry other)
Liquid
Clear colorless to pale yellow
15, 1648
99%
Calcium
99%
Ca
40.07
100g
05,89; 09,1634; 14,71
1 % max.
7440-70-2
Ca
Glass bottle
MFCD00085314

Chemical Identifiers

7440-70-2
99%
Ca
MFCD00085314
≥98.5% (Titrimetry other)
Liquid
Ca
40.07
Calcium, 99%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
In contact with water releases flammable gases.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Handle under inert gas. Protect from moisture.
Keep away from any possible contact with water, because of violent reaction and possible flash fire.
Brush off loose particles from skin. Immerse in cool water/wrap in wet bandages.
In case of fire: Use CO2, dry chemical, or foam.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger