Learn More
Calcium, 99%, granular, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 201185000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Gray | |
99% | |
7440-70-2 | |
99% | |
Ca | |
500 g | |
15, 1648 | |
OYPRJOBELJOOCE-UHFFFAOYSA-N | |
calcium | |
5460341 | |
40.07 | |
Plastic Bottle |
Calcium | |
845.0°C | |
1484.0°C | |
Ca | |
MFCD00010897 MFCD00085314 | |
05,89; 09,1634; 14,71 | |
unii-sy7q814vup, hsdb 273, sy7q814vup, compounds, elemental, calcium, elemental, kalzium, atom, ingot, chelate | |
[Ca] | |
40.08 | |
CHEBI:29320 | |
99% | |
Granular |
Chemical Identifiers
7440-70-2 | |
40.08 | |
OYPRJOBELJOOCE-UHFFFAOYSA-N | |
5460341 | |
calcium |
Ca | |
MFCD00010897 MFCD00085314 | |
unii-sy7q814vup, hsdb 273, sy7q814vup, compounds, elemental, calcium, elemental, kalzium, atom, ingot, chelate | |
CHEBI:29320 | |
[Ca] |
Safety and Handling
GHS H Statement:
In contact with water releases flammable gases.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Handle under inert gas. Protect from moisture.
Keep away from any possible contact with water, because of violent reaction and possible flash fire.
Brush off loose particles from skin. Immerse in cool water/wrap in wet bandages.
In case of fire: Use CO2, dry chemical, or foam.
Store in a dry place. Store in a closed container.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 231-179-5
TSCA : TSCA
RUO – Research Use Only