missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calcium, 99%, granular, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC201180050

 View more versions of this product

Catalog No. AC201180050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Calcium
99%
99%
Ca
05,89; 09,1634; 14,71
atom, calcium, elemental, chelate, compounds, elemental, hsdb 273, ingot, kalzium, sy7q814vup, unii-sy7q814vup
[Ca]
40.078
5460341
40.07
99%
845.0°C
5g
7440-70-2
Ca
MFCD00085314
15, 1648
OYPRJOBELJOOCE-UHFFFAOYSA-N
calcium
Glass bottle
CHEBI:29320
Granules
Gray
1484.0°C

Chemical Identifiers

7440-70-2
40.078
OYPRJOBELJOOCE-UHFFFAOYSA-N
5460341
calcium
Ca
MFCD00085314
atom, calcium, elemental, chelate, compounds, elemental, hsdb 273, ingot, kalzium, sy7q814vup, unii-sy7q814vup
CHEBI:29320
[Ca]
Safety and Handling

Safety and Handling

GHS H Statement:
In contact with water releases flammable gases.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Handle under inert gas. Protect from moisture.
Keep away from any possible contact with water, because of violent reaction and possible flash fire.
Brush off loose particles from skin. Immerse in cool water/wrap in wet bandages.
In case of fire: Use CO2, dry chemical, or foam.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 231-179-5

TSCA : TSCA

SDS Documents