missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Calcium 4-Methyl-2-oxovalerate Hydrate 98.0+%, TCI America™
Supplier: TCI America K002325G
Specifications
Calcium 4-Methyl-2-oxovalerate Hydrate | |
White | |
MFCD00004163 | |
4-Methyl-2-oxovaleric Acid Calcium Salt | |
CC(C)CC(=O)C(=O)O.CC(C)CC(=O)C(=O)O.[Ca] | |
300.364 | |
298.35 | |
Crystalline Powder |
51828-95-6 | |
C12H20CaO6 | |
25 g | |
QSTGQEJKDFGVPP-UHFFFAOYSA-N | |
calcium;4-methyl-2-oxopentanoic acid | |
131854617 | |
≥98.0% (T) |
Chemical Identifiers
51828-95-6 | |
300.364 | |
QSTGQEJKDFGVPP-UHFFFAOYSA-N | |
131854617 | |
CC(C)CC(=O)C(=O)O.CC(C)CC(=O)C(=O)O.[Ca] |
C12H20CaO6 | |
MFCD00004163 | |
4-Methyl-2-oxovaleric Acid Calcium Salt | |
calcium;4-methyl-2-oxopentanoic acid |
Safety and Handling
TSCA : No