missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Calcium 2,4-pentanedionate hydrate, 99%

Manufacturer:  Alfa Aesar™ 3052618

 View more versions of this product

Catalog No. AA3052618



Specifications

Specifications

Calcium 2,4-pentanedionate hydrate
99%
MFCD00013486
FFWWBMKQRYMOKA-SUKNRPLKSA-L
calcium;(Z)-4-oxopent-2-en-2-olate;hydrate
16219078
Crystalline
50g
345909-31-1
C10H16CaO5
Calcium acetylacetonate
CC(=CC(=O)C)[O-].CC(=CC(=O)C)[O-].O.[Ca+2]
256.311
238.30 (Anhydrous)
175°C (decomposition)

Chemical Identifiers

345909-31-1
256.311
FFWWBMKQRYMOKA-SUKNRPLKSA-L
16219078
CC(=CC(=O)C)[O-].CC(=CC(=O)C)[O-].O.[Ca+2]
C10H16CaO5
MFCD00013486
Calcium acetylacetonate
calcium;(Z)-4-oxopent-2-en-2-olate;hydrate
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents