Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Calcitonin, Rabbit Monoclonal Antibody

Identify a spectrum of C cell proliferative abnormalities ranging from C cell hyperplasia to invasive tumors using Thermo Scientific™ Calcitonin Rabbit Monoclonal Antibody.

$305.69 - $1,270.08

Specifications

Antigen Calcitonin
Clone SP17
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

RM9117R7

 
thermo scientific™ lab vision™
RM9117R7
7mL Each for $647.86

RM9117S

 
thermo scientific™ lab vision™
RM9117S
1mL Each for $1,270.08

RM9117S0

 
thermo scientific™ lab vision™
RM9117S0
100µL Each for $305.69

RM9117S1

 
thermo scientific™ lab vision™
RM9117S1
500µL Each for $817.82
Description & Specifications

Specifications

Antigen Calcitonin
Clone SP17
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
Host Species Rabbit
Regulatory Status IVD
Research Discipline Steroids Hormones and Growth Factors
Type Primary
Immunogen Synthetic human calcitonin 1-32 amino acid peptide

Identify a spectrum of C cell proliferative abnormalities ranging from C cell hyperplasia to invasive tumors using Thermo Scientific™ Calcitonin, Rabbit Monoclonal Antibody. Studies show rabbit monoclonal antibodies are more sensitive and specific than other available markers.

Calcitonin is a 32 amino acid peptide which can be demonstrated in C cells of the normal and hyperplastic thyroid. Staining for calcitonin may be used for the identification of a spectrum of C cell proliferative abnormalities ranging from C cell hyperplasia to invasive tumors. Staining for calcitonin in medullary carcinoma of the thyroid produces a fine granular pattern in the cytoplasm. Amyloid deposits within the tumor may also exhibit varying degrees of calcitonin activity.

Host Species: Rabbit

Clone: SP17

Speices Reactivity: Human, Monkey, Mouse, Rat, Dog, Horse, Sheep and Pig. Others not tested.

Epitope: 1-32aa

Immunogen: Synthetic human calcitonin 1-32 amino acid peptide

Positive Control: Thyroid or medullary CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)