missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific Chemicals Calcitonin Gene Related Peptide, Rat
For cellular and molecular biology applications
Supplier: Thermo Scientific Chemicals J66827EXD
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Rat | |
-30°C to -10°C | |
3806.24 | |
0.5 mg | |
CGRP, rat |
HPLC | |
Solid | |
Calcitonin Gene Related Peptide, rat | |
RUO - research use only |
RUO – Research Use Only