Please login to your online account to display your discounted pricing

Alfa Aesar™ Calcitonin Gene Related Peptide, Rat


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J66827MCR

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ66827MCR

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Product Type Calcitonin Gene Related Peptide, rat
Quantity 1mg
Storage Requirements -30° to -10°C
Molecular Weight 3806.24
Cross Reactivity Rat
Sequence CGRP, rat
Purification Method HPLC
Regulatory Status RUO
Format Solid