Please login to your online account to display your discounted pricing

Alfa Aesar™ Calcitonin Gene Related Peptide (8-37), Human

For cellular and molecular biology applications

Specifications

Product Type Calcitonin Gene Related Peptide (8-37), human
Cross Reactivity Human
Molecular Weight 3125.58
Storage Requirements -30° to -10°C
Sequence α-CGRP (8-37), human
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ65912EXD

 
alfa aesar™
J65912EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ65912MCR

 
alfa aesar™
J65912MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Calcitonin Gene Related Peptide (8-37), human
Cross Reactivity Human
Molecular Weight 3125.58
Storage Requirements -30° to -10°C
Sequence α-CGRP (8-37), human
Format Powder
Regulatory Status RUO
Purification Method HPLC