Alfa Aesar™ Calcitonin Gene Related Peptide (8-37), Human
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar™ J65912MCR
 View more versions of this product
Catalog No. AAJ65912MCR
Description
Description
Specifications
Specifications
3125.58
Human
Powder
RUO
Calcitonin Gene Related Peptide (8-37), human
HPLC
1mg
α-CGRP (8-37), human
-30° to -10°C
Powder