missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Calcitonin Ab-2, Rabbit Polyclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ Calcitonin Ab-2, Rabbit Polyclonal Antibody.

Manufacturer:  Thermo Scientific™ Lab Vision™ RB1519PABX

 View more versions of this product

Catalog No. RB1519PABX


 

Specifications

Description

Ensure accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ Calcitonin Ab-2, Rabbit Polyclonal Antibody.

Calcitonin is demonstrated in C cells of the normal and hyperplastic thyroid. Staining for calcitonin may be used for the identification of a spectrum of C cell proliferative abnormalities ranging from C cell hyperplasia to invasive tumors. Staining for calcitonin in medullary carcinoma of the thyroid produces a fine granular pattern in the cytoplasm.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Monkey, Mouse, Rat, Dog, Horse, Sheep and Pig. Others not tested.

Immunogen: A synthetic peptide from human calcitonin

Positive Control: Thyroid or medullary CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

Calcitonin Ab-2
Polyclonal
Unlabeled
Concentrated
A synthetic peptide from human calcitonin
1mL
Steroids Hormones and Growth Factors
Polyclonal
Immunohistochemistry (Paraffin)
1mg/mL
Ab without BSA/Azide (Frozen)
Rabbit
IgG
RUO
Primary
Canine,Equine,Human,Murine,Non-human Primate,Porcine,Rat