Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Calcitonin Ab-2, Rabbit Polyclonal Antibody


Ensure accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ Calcitonin Ab-2, Rabbit Polyclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  RB1519P1ABX

 View more versions of this product

Catalog No. RB1519P1ABX

  • / Each
 

Description & Specifications

Specifications

Host Species Rabbit
Cross Reactivity Canine
Cross Reactivity Equine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Non-Human Primate
Cross Reactivity Porcine
Cross Reactivity Rat
Antigen Calcitonin Ab-2
Immunogen A synthetic peptide from human calcitonin
Applications Immunohistochemistry (Paraffin)
Quantity 500µL
Format Concentrated
Regulatory Status RUO
Isotype IgG
Formulation Ab without BSA/Azide (Frozen)
Research Discipline Steroids Hormones and Growth Factors
Conjugate Unlabeled
Classification Polyclonal
Type Primary
Concentration 1mg/mL
Item Description 500uL; 1mg/mL; Purified w/o BSA, azide

Ensure accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ Calcitonin Ab-2, Rabbit Polyclonal Antibody.

Calcitonin is demonstrated in C cells of the normal and hyperplastic thyroid. Staining for calcitonin may be used for the identification of a spectrum of C cell proliferative abnormalities ranging from C cell hyperplasia to invasive tumors. Staining for calcitonin in medullary carcinoma of the thyroid produces a fine granular pattern in the cytoplasm.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Monkey, Mouse, Rat, Dog, Horse, Sheep and Pig. Others not tested.

Immunogen: A synthetic peptide from human calcitonin

Positive Control: Thyroid or medullary CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO