Please login to your online account to display your discounted pricing

Alfa Aesar™ Calcitonin (8-32), Salmon

For cellular and molecular biology applications

Specifications

Product Type Calcitonin (8-32), salmon
Cross Reactivity Salmon
Molecular Weight 2725.06
Storage Requirements -30° to -10°C
Sequence Val-Leu-Gly-Lys-Leu-Ser-Gln-Glu-Leu-His-Lys-Leu-Gln-Thr-Tyr-Pro-Arg-Thr-Asn-Thr-Gly-Ser-Gly-Thr-Pro-NH2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ64415MCR

 
alfa aesar™
J64415MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Calcitonin (8-32), salmon
Cross Reactivity Salmon
Molecular Weight 2725.06
Storage Requirements -30° to -10°C
Sequence Val-Leu-Gly-Lys-Leu-Ser-Gln-Glu-Leu-His-Lys-Leu-Gln-Thr-Tyr-Pro-Arg-Thr-Asn-Thr-Gly-Ser-Gly-Thr-Pro-NH2
Format Solid
Regulatory Status RUO
Purification Method HPLC