Please login to your online account to display your discounted pricing

Calcipotriol, Tocris

Vitamin D3 analog

$246.52 - $1,023.37

Chemical Identifiers

Chemical Name or Material Calcipotriol
Assay Percent Range >98%
Molecular Formula C27H40O3
Formula Weight 412.6
Synonym (1α,3β,5Z,7E,22E,24S)-24-Cyclopropyl-9,10-secochola-5,7,10(19),22-tetraene-1,3,24-triol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

270050

 
r & d systems
RDS270050
50mg Each for $1,023.37

270010

 
r & d systems
RDS270010
10mg Each for $246.52
Description & Specifications

Specifications

Formula Weight 412.6
Quantity 50mg
Chemical Name or Material Calcipotriol
Synonym (1α,3β,5Z,7E,22E,24S)-24-Cyclopropyl-9,10-secochola-5,7,10(19),22-tetraene-1,3,24-triol
CAS 112965-21-6
Assay Percent Range >98%
Molecular Formula C27H40O3
Recommended Storage Desiccate at -20°C
Safety and Handling

Desiccate at -20°C