missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calciferol, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals J6161003

 View more versions of this product

Catalog No. AAJ6161003



Description

Description

Calciferol is a strong inhibitory effect against bladder tumor promotion by sodium saccharin and induces cell differentiation in leukemia cells. Calciferol also been shown to be an inhibitor of DNA polymerase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Calciferol is a strong inhibitory effect against bladder tumor promotion by sodium saccharin and induces cell differentiation in leukemia cells. Calciferol also been shown to be an inhibitor of DNA polymerase.

Solubility
Soluble in ethanol, water, methanol, dimethylformamide, and dimethyl sulfoxide.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

Calciferol
114°C to 118°C
C28H44O
1 g
1916682
ergocalciferol, vitamin d2, calciferol, viosterol, oleovitamin d2, ercalciol, ergorone, detalup, condocaps, crystallina
MECHNRXZTMCUDQ-VLOQVYPSSA-N
(1S)-3-[2-[(1S,7aR)-1-[(2R,5R)-5,6-dimethylhept-3-en-2-yl]-7a-methyl-2,3,3a,5,6,7-hexahydro-1H-inden-4-ylidene]ethylidene]-4-methylidenecyclohexan-1-ol
134129658
Powder
50-14-6
White
MFCD00166988,MFCD11656674
UN2811
14,10018
Soluble in ethanol,water,methanol,dimethylformamide,and dimethyl sulfoxide.
CC(C)[C@@H](C)\C=C\[C@@H](C)[C@H]1CC[C@H]2\C(CCC[C@]12C)=C\C=C1/C[C@@H](O)CCC1=C
396.66
396.65

Chemical Identifiers

50-14-6
396.66
MECHNRXZTMCUDQ-VLOQVYPSSA-N
134129658
CC(C)[C@@H](C)\C=C\[C@@H](C)[C@H]1CC[C@H]2\C(CCC[C@]12C)=C\C=C1/C[C@@H](O)CCC1=C
C28H44O
MFCD00166988,MFCD11656674
ergocalciferol, vitamin d2, calciferol, viosterol, oleovitamin d2, ercalciol, ergorone, detalup, condocaps, crystallina
(1S)-3-[2-[(1S,7aR)-1-[(2R,5R)-5,6-dimethylhept-3-en-2-yl]-7a-methyl-2,3,3a,5,6,7-hexahydro-1H-inden-4-ylidene]ethylidene]-4-methylidenecyclohexan-1-ol
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H372
Fatal if swallowed.
Fatal in contact with skin.
Causes damage to organs through prolonged or repeated exposure.

P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P310-P312-P314-P330-P361-P363-P501c

H301+H311-H330-H372

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 200-014-9

RTECSNumber : KE1050000

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only