missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Calcein sodium salt, ca 2-3 Na

$72.04 - $223.19

Chemical Identifiers

CAS 108750-13-6
Molecular Formula C30H24N2Na2O13
Formula Weight 666.51
MDL Number MFCD00005049
Synonym Bis[N,N-bis(carboxymethyl)aminomethyl]fluorescein sodium salt; Fluorexon
Chemical Name or Material Calcein sodium salt, ≈2 to 3 Na
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1025506 SDS Alfa Aesar™
L1025506
5g Each for $72.04
AAL1025514 SDS Alfa Aesar™
L1025514
25g Each for $223.19
Specifications

Description

Applications
Calcein sodium salt is used as an indicator for the complexometric determination of calcium and magnesium. It is also used as an indicator for the determination of calcium in biological systems.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Description & Specifications

Chemical Identifiers

108750-13-6
666.51
Bis[N,N-bis(carboxymethyl)aminomethyl]fluorescein sodium salt; Fluorexon
C30H24N2Na2O13
MFCD00005049
Calcein sodium salt, ≈2 to 3 Na

Specifications

5g
C30H24N2Na2O13
MFCD00005049
Calcein sodium salt, ≈2 to 3 Na
108750-13-6
666.51
Bis[N,N-bis(carboxymethyl)aminomethyl]fluorescein sodium salt; Fluorexon