missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Honeywell Fluka™ Calcein Disodium Salt, Honeywell™ Fluka™

for complexometry

Manufacturer:  Honeywell Fluka™ 210301G

 View more versions of this product

Catalog No. 60045202



Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Specifications

Calcein Disodium Salt
108750-13-6
C30H25N2NaO13
NONH for all modes of transport
C1=CC=C2C(=C1)C(=O)OC23C4=CC(=C(C=C4OC5=CC(=C(C=C35)CN(CC(=O)O)CC(=O)O)[O-])O)CN(CC(=O)O)CC(=O)O.[Na+]
644.521
666.5g/mol
Red
For complexometry
Powder
MFCD00082336
WFTKXUHBPZFOMD-UHFFFAOYSA-M
sodium;2',7'-bis[[bis(carboxymethyl)amino]methyl]-6'-hydroxy-3-oxospiro[2-benzofuran-1,9'-xanthene]-3'-olate
131674686
Glass Bottle
1g

Chemical Identifiers

108750-13-6
644.521
WFTKXUHBPZFOMD-UHFFFAOYSA-M
sodium;2',7'-bis[[bis(carboxymethyl)amino]methyl]-6'-hydroxy-3-oxospiro[2-benzofuran-1,9'-xanthene]-3'-olate
C30H25N2NaO13
MFCD00082336
131674686
C1=CC=C2C(=C1)C(=O)OC23C4=CC(=C(C=C4OC5=CC(=C(C=C35)CN(CC(=O)O)CC(=O)O)[O-])O)CN(CC(=O)O)CC(=O)O.[Na+]
Documents