Please login to your online account to display your discounted pricing

EMD Millipore™ Calbiochem™ Uridine

For biochemical applications

$107.26 - $293.31

Specifications

Product Type Uridine
Storage Requirements 15° to 30°C
Format White to off-white solid
Purity ≥97% by HPLC; ≥99% by UV
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

6680100GM

 
emd millipore™
6680100GM
100g Each for $293.31

668025GM

 
emd millipore™
668025GM
25g Each for $107.26
Description & Specifications

Specifications

Product Type Uridine
Storage Requirements 15° to 30°C
Format White to off-white solid
Purity ≥97% by HPLC; ≥99% by UV