MilliporeSigma™ Calbiochem™ Thyroid Stimulating Hormone, Human Pituitary, Iodination Grade
Glycoprotein hormone that exerts mild, continuous stimulation on thyroid, resulting in maintenance of activity
Supplier: MilliporeSigma™ 86900610UG
Description
- Secretion is inhibited by Somatostatin
Specifications
10μg | |
-20°C | |
Thyroid Stimulating Hormone, Human Pituitary | |
≥95% by weight |
Prepared from tissue of individuals that have been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV. | |
Thyrotropin, hTSH | |
Lyophilized |
For Research Use Only