missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MilliporeSigma™ Calbiochem™ Thyroid Stimulating Hormone, Human Pituitary, Iodination Grade
Glycoprotein hormone that exerts mild, continuous stimulation on thyroid, resulting in maintenance of activity
$280.00 - $1330.00
Specifications
Source | Prepared from tissue of individuals that have been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV. |
---|---|
Storage Requirements | -20°C |
Common Name | Thyrotropin, hTSH |
Product Type | Thyroid Stimulating Hormone, Human Pituitary |
Form | Lyophilized |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
86900610UG
|
MilliporeSigma™
86900610UG |
10μg |
Each for $280.00
|
|
|||||
86900650UG
|
MilliporeSigma™
86900650UG |
50μg |
Each for $779.00
|
|
|||||
869006100UG
|
MilliporeSigma™
869006100UG |
100μg |
Each for $1,330.00
|
|
|||||
Description
- Secretion is inhibited by Somatostatin
Specifications
Prepared from tissue of individuals that have been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV. | |
Thyrotropin, hTSH | |
Lyophilized |
-20°C | |
Thyroid Stimulating Hormone, Human Pituitary | |
≥95% by weight |
For Research Use Only