Please login to your online account to display your discounted pricing

EMD Millipore Calbiochem™ S1P1 Receptor Agonist, SEW2871

The S1P1 Receptor Agonist, SEW2871, controls the biological activity of S1P1 Receptor. This small molecule/inhibitor is primarily used for Activators/Inducers applications.

$281.14 - $711.47

Specifications

Storage Requirements Protect from Light
Molecular Formula C20H10F6N2OS
Format White solid
Formula Weight 440.4g/mol
Target Proteins S1P1 Receptor
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

56-773-31MG

 
emd millipore
5677331MG
1mg Each for $281.14

56-773-35MG

 
emd millipore
5677335MG
5mg Each for $711.47
Description & Specifications

Specifications

Storage Requirements Protect from Light
Molecular Formula C20H10F6N2OS
Format White solid
Formula Weight 440.4g/mol
Target Proteins S1P1 Receptor
Description 98% by HPLC
Solubility DMSO

  • Packaged Under Inert Gas
  • Cell Permeable