MilliporeSigma™ Calbiochem™ Purmorphamine
A cell-permeable activator of Hedgehog signaling that induces osteoblast differentiation of multipotent mesenchymal progenitor cells C3H10T1/2 (EC50 = 1 μM).
Supplier: MilliporeSigma™ 5402205MG
Description
- Packaged Under Inert Gas
- Cell Permeable
Specifications
Off-white | |
Activation of the Hedgehog signaling pathway | |
C31H32N6O2 | |
Protect from light | |
5 mg |
98% by HPLC | |
483367-10-8 | |
DMSO | |
Solid |