Please login to your online account to display your discounted pricing

EMD Millipore Calbiochem™ Protease Inhibitor Cocktail Set VII

Protease Inhibitor Cocktail Set VII contains five protease inhibitors with broad specificity. Recommended for purification of proteins containing His·Tag® sequences.

$82.00 - $392.00

Specifications

Format Liquid
Formulation In 1mL DMSO.
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

53-913-81ML

 
emd millipore
5391381ML
1mL Each for $82.00

53-913-81SET

 
emd millipore
5391381SET
1 set Each for $392.00
Description & Specifications

Specifications

Format Liquid
Formulation In 1mL DMSO.