Please login to your online account to display your discounted pricing

EMD Millipore Calbiochem™ Protease Inhibitor Cocktail Set III, EDTA-Free

Protease inhibitor cocktail III, EDTA-free for inhibiting aspartic, cysteine, and serine proteases as well as aminopeptidases in mammalian cells and tissues.

$102.00 - $102.00

Specifications

Format Liquid
Target Proteins Aspartic, cysteine, and serine proteases as well as aminopeptidases
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

53-913-41ML

 
emd millipore
5391341ML
1mL Each for $102.00
Description & Specifications

Specifications

Format Liquid
Target Proteins Aspartic, cysteine, and serine proteases as well as aminopeptidases