MilliporeSigma™ Calbiochem™ NADPH, Tetrasodium Salt
Ubiquitous coenzyme that acts as an electron donor in many reactions utilizing dehydrogenase and reductase enzymes
Supplier: MilliporeSigma™ 481973500MG
Specifications
-20°C | |
NADPH, Tetrasodium Salt | |
500 mg |