Please login to your online account to display your discounted pricing

EMD Millipore™ Calbiochem™ Laminin Nonapeptide


Active in cell adhesion and cell-surface receptor binding and in stimulating cell migration

Manufacturer: emd millipore™  052337035MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. 052337035MG

  • / Each
 

Description & Specifications

Specifications

Format White to off-white lyophilized solid
Sequence H-Cys-Asp-Pro-Gly-Tyr-Ile-Gly-Ser-Arg-OH
Purity ≥90% by HPLC
Storage Requirements -20°C
Product Type Laminin Nonapeptide
Quantity 5mg