missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

EMD Millipore™ Calbiochem™ Human Angiotensin II

Plays important role in regulation of blood pressure

$67.78 - $294.06

Specifications

Product Type Human Angiotensin II
Format White to off-white solid
Purity ≥98% by HPLC
Storage Requirements -20°C
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
052301011MG EMD Millipore™
052301011MG
1mg Each for $67.78
05-230-10125M EMD Millipore™
0523010125MG
25mg Each for $294.06
052301015MG EMD Millipore™
052301015MG
5mg Each for $85.36
Specifications

Description

  • Stimulates release of aldosterone from adrenal gland
  • Inhibits adenylate cyclase activity in spontaneously hypertensive rats
  • Has strong vasoconstrictive effects
  • Increases the entry of Ca2+ in heart muscle via voltage-sensitive channels
  • Activates myosin light chain kinase
  • Elicits increase in voltage-dependent delayed rectifier K+ current in neurons
  • Activates JAK2 in smooth muscle cells
Description & Specifications

Specifications

Human Angiotensin II
White to off-white solid
≥98% by HPLC
-20°C