Please login to your online account to display your discounted pricing

EMD Millipore™ Calbiochem™ Human Angiotensin II


Plays important role in regulation of blood pressure

Manufacturer: emd millipore™  0523010125MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. 05-230-10125M

  • / Each
 

Description & Specifications

Specifications

Product Type Human Angiotensin II
Format White to off-white solid
Quantity 25mg
Storage Requirements -20°C
Purity ≥98% by HPLC