missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ High Purity IPTG, Animal-Free

Used in stimulation of β-galactosidase in cellular systems in which dioxane would disrupt normal cell function

$141.00 - $355.00

Specifications

Content And Storage 2- 8°C
Form White to off-white solid
Product Type IPTG
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Qty  
Catalog Number Mfr. No. Quantity Price Qty  
4202911GM
View Documents
MilliporeSigma™
4202911GM
1 g
Each for $141.00
Only null left
 
4202915GM
View Documents
MilliporeSigma™
4202915GM
5 g
Each for $355.00
Only null left
 
Description

Description

  • Contains no animal-derived products
  • Inducer of β-D-galactosidase
Specifications

Specifications

2- 8°C
White to off-white solid
IPTG
Videos
SDS
Documents

Documents

For Research Use Only