Please login to your online account to display your discounted pricing

EMD Millipore™ Calbiochem™ H-Gly-Arg-Ala-Asp-Ser-Pro-OH


Inactive control peptide for other fibronectin inhibitors

Manufacturer: emd millipore™  0334005225M

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. 0334005225M

  • / Each
 

Description & Specifications

Specifications

Storage Requirements -20°C
Quantity 25mg
Product Type H-Gly-Arg-Ala-Asp-Ser-Pro-OH
Formulation Supplied as a trifluoroacetate salt
Sequence H-Gly-Arg-Ala-Asp-Ser-Pro-OH
Format White to off-white lyophilized solid
Purity ≥98% by HPLC