Please login to your online account to display your discounted pricing

EMD Millipore™ Calbiochem™ FURA 2/AM

Cell-permeable ester form of FURA 2

$215.47 - $215.47

Specifications

CAS 108964-32-5
Color Yellow
Format Solid
Formula Weight 1001.9g/mol
Packaging Type Packaged under inert gas
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

3449051MG

 
emd millipore™
3449051MG
1mg Each for $215.47
Description & Specifications

Specifications

CAS 108964-32-5
Color Yellow
Format Solid
Formula Weight 1001.9g/mol
Packaging Type Packaged under inert gas
Purity ≥95% by HPLC
Storage Requirements Store at −20°C
Synonym 1-[2-(5-Carboxyoxazol-2-yl)-6-aminobenzofuran-5-oxy]-2-(2′-amino-5′-methylphenoxy)-ethane-N,N,N′N′-tetraacetic Acid Pentaacetoxymethyl Ester

  • Cell-permeable ester form of FURA 2
  • PLURONIC™ F-127, PROTEIN GRADE™ Detergent is a mild, low-toxicity detergent that can aid in the solubilization of FURA 2/AM