Please login to your online account to display your discounted pricing

EMD Millipore™ Calbiochem™ BQ-123, Sodium Salt


Highly potent, selective, and competitive ETA receptor antagonist

Manufacturer: emd millipore™  052338315MG

 View more versions of this product

Catalog No. 052338315MG

  • / Each
 

Description & Specifications

Specifications

Quantity 1mg
Format Solid
Storage Requirements Store at −20°C
Formula Weight 632.7g/mol
Color White
Molecular Formula C31H41N6O7 · Na
Formulation Supplied as a sodium salt. Sold on the basis of peptide content
Target ETA (endothelin) receptor antagonist
Purity ≥95% by HPLC

Primary target Ki: 40nM for ETA versus 2.5μM for ETB