Please login to your online account to display your discounted pricing

EMD Millipore™ Calbiochem™ Aquacide III

Useful for concentration of proteins and other macromolecules and can be used for rapid removal of water from dialysis tubing

$107.26 - $392.03

Specifications

CAS 25322-68-3
Color Colorless
Format Flakes
Formula Weight 8000g/mol
Storage Requirements Store at +15°C to +30°C
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

178521KG

 
emd millipore™
178521KG
1kg Each for $107.26

178525KG

 
emd millipore™
178525KG
5kg Each for $392.03
Description & Specifications

Specifications

CAS 25322-68-3
Color Colorless
Format Flakes
Formula Weight 8000g/mol
Storage Requirements Store at +15°C to +30°C

  • Useful for concentration of proteins and other macromolecules and can be used for rapid removal of water from dialysis tubing by using either a 2% syrup or the Aquacide solid
  • Neither denature proteins nor change the pH during concentration
  • At room temperature 1g of Aquacide absorbs about 5mL of water from solution in 1h