Please login to your online account to display your discounted pricing

EMD Millipore Calbiochem™ Aprotinin, Bovine Lung, Solution

Aprotinin, Bovine Lung, Solution, CAS 9087-70-1, is a competitive, reversible, heat and acid-stable inhibitor of proteolytic and esterolytic activities.

$834.38 - $834.38

Specifications

Format Liquid
Formulation In isotonic NaCl, aseptically filled.
Formula Weight 6512g/mol
Target Proteins chymotrypsin
Concentration 6,500 KIU/mL
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

616399500KU

 
emd millipore
616399500KU
500 ku Each for $834.38
Description & Specifications

Specifications

Format Liquid
Formulation In isotonic NaCl, aseptically filled.
Formula Weight 6512g/mol
Target Proteins chymotrypsin
Concentration 6,500 KIU/mL