missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MilliporeSigma™ Calbiochem™ Aprotinin, Bovine Lung, Crystalline
Aprotinin, Bovine Lung, Crystalline, CAS 9087-70-1, is a competitive, reversible, heat- and acid-stable inhibitor of proteolytic and esterolytic activities.
Supplier: MilliporeSigma™ 616370100MG
Description
Contaminants: endotoxin: ≤10 EU/mgSpecifications
5500 KIU/mg, dry weight | |
95% by gel filtration | |
9087-70-1 | |
H2O | |
Crystalline Solid |
White to Off-white | |
chymotrypsin | |
6512g/mol | |
Hygroscopic | |
100 mg |