Please login to your online account to display your discounted pricing

EMD Millipore™ Calbiochem™ Ampicillin, Sodium Salt

Suitable for use in research that uses ampicillin-resistant plasmids

$69.83 - $234.46

Specifications

Product Type Ampicillin
Storage Requirements 2- 8°C
Format White solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

17125425GM

 
emd millipore™
17125425GM
25g Each for $234.46

1712545GM

 
emd millipore™
1712545GM
5g Each for $69.83
Description & Specifications

Specifications

Product Type Ampicillin
Storage Requirements 2- 8°C
Format White solid

  • β-lactam inhibits bacterial cell-wall synthesis
  • Active against Gram-negative bacteria
  • Inactivated by lactamases