MilliporeSigma™ Calbiochem™ Protease Inhibitor Cocktail Set III, EDTA-Free
Protease inhibitor cocktail III, EDTA-free for inhibiting aspartic, cysteine, and serine proteases as well as aminopeptidases in mammalian cells and tissues.
Supplier: MilliporeSigma™ 5391341ML
Description
A cocktail of six protease inhibitors with broad specificity for the inhibition of aspartic, cysteine, and serine proteases as well as aminopeptidases. This cocktail is recommended for use with mammalian cell and tissue extracts, but is also suitable for bacterial extracts. Supplied with a data sheet.Specifications
Aspartic, cysteine, and serine proteases as well as aminopeptidases | |
Liquid | |
1 mL |