Please login to your online account to display your discounted pricing

EMD Millipore™ Calbiochem™ Adenosine 5′-Triphosphate, Disodium Salt

For biochemical applications

$46.27 - $470.83

Specifications

Product Type Adenosine 5′-Triphosphate, Disodium Salt
Storage Requirements -20°C
Format White solid
Purity ≥95% by HPLC
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

11911GM

 
emd millipore™
11911GM
1g Each for $46.27

119125GM

 
emd millipore™
119125GM
25g Each for $274.33

119150GM

 
emd millipore™
119150GM
50g Each for $470.83

11915GM

 
emd millipore™
11915GM
5g Each for $69.83
Description & Specifications

Specifications

Product Type Adenosine 5′-Triphosphate, Disodium Salt
Storage Requirements -20°C
Format White solid
Purity ≥95% by HPLC