missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

EMD Millipore™ Calbiochem™ Adenosine 5′-Diphosphate, Monosodium Salt

For biochemical applications

$107.26 - $323.70

Specifications

Product Type Adenosine 5′-Diphosphate, Monosodium Salt
Storage Requirements 2° to 8°c
Format White solid
Purity ≥95% by HPLC
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
11711GM EMD Millipore™
11711GM
1g Each for $107.26
11715GM EMD Millipore™
11715GM
5g Each for $323.70
Specifications Description & Specifications

Specifications

Adenosine 5′-Diphosphate, Monosodium Salt
2° to 8°c
White solid
≥95% by HPLC