missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calapro Powder-Free Latex Finger Cots

Made of 100% pure, super-thin latex (5mil thick) for comfort and dexterity

Manufacturer:  Calapro Inc FY01S

 View more versions of this product

Catalog No. 11-392-9A



 

Alert:

These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.


Specifications Description & Specifications

Specifications

5mil
7cm
0.62 in.
Finger Cot
For comfort and dexterity
Natural Latex
Yes
2.75 in.
ISO class 5
Small
Ember
1.6cm
4.5