missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calapro Powder-Free Latex Finger Cots

Made of 100% pure, super-thin latex (5mil thick) for comfort and dexterity

$44.16 - $44.16

Specifications

Certifications/Compliance ISO class 5
Color Ember
Product Type Finger Cot
For Use With (Application) For comfort and dexterity
Length (English) 2.75 in.
View More Specs

Alert:

These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.



Products ${productFamilyLength}
Catalog Number Mfr. No. Diameter (English) Diameter (Metric) Size Price Quantity  
Catalog Number Mfr. No. Diameter (English) Diameter (Metric) Size Price Quantity  
11-392-9A Calapro Inc
FY01S
0.62 in. 1.6cm Small Pack of 1440 Each for $44.16
Case of 20 Pack for $846.00
11-392-9C Calapro Inc
FY01L
0.78 in. 2cm Large Pack of 1440 Each for $44.16
Case of 20 Pack for $846.00
11-392-9D Calapro Inc
FY01X
0.86 in. 2.2cm XL Pack of 1440 Each for $44.16
Case of 20 Pack for $846.00
11-392-9B Calapro Inc
FY01M
0.70 in. 1.8cm Medium Pack of 1440 Each for $44.16
Case of 20 Pack for $846.00
Specifications

Description

Finger cots fit without bulkiness at the tip to reduce finger fatigue.

  • Chlorination-washed to minimize contamination
  • Length: 2 13/16 in. (7cm)

Order Info

Rolled, 1140 cots to a pack.

Compliance

Inspected and packaged under ISO Class 5 (formerly Class 100) cleanroom specifications

Description & Specifications

Specifications

ISO class 5
Finger Cot
2.75 in.
Natural Latex
4.5
Ember
For comfort and dexterity
7cm
Yes
5mil