Please login to your online account to display your discounted pricing

Calapro Antistatic Latex Finger Cots

Antistatic, super-thin.

$30.15 - $30.15

Specifications

Certifications/Compliance ISO Class 5
Color Pink
Product Type Antistatic Finger Cot
For Use With (Application) For static-free areas
For Use With (Equipment) Used when handling Class II category B (=1000V) static-sensitive devices
View More Specs

 Alert

Warning: The products featured on this page contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.

Products
Catalog Number Mfr. No. Material Size Price Quantity    

11-392-16B

 
calapro inc
FW04M
Natural Rubber Medium Pack of 720 Pair for $30.15
Case of 40 Pack for $1,155.20

11-392-16C

 
calapro inc
FW04L
Natural Rubber latex Large Pack of 720 Pair for $30.15
Case of 40 Pack for $1,155.20

11-392-16D

 
calapro inc
FW04X
Natural Rubber latex XL Pack of 720 Each for $30.15
Case of 40 Pack for $1,155.20
Description & Specifications

Specifications

Certifications/Compliance ISO Class 5
Color Pink
Product Type Antistatic Finger Cot
For Use With (Application) For static-free areas
For Use With (Equipment) Used when handling Class II category B (=1000V) static-sensitive devices
Length (English) 2.75 in.
Length (Metric) 7cm
Powder-Free Yes
Thickness (English) 5mil

Made of 100% pure, super-thin latex (5mil thick) for comfort and dexterity. Fit without bulkiness at the tip to reduce finger fatigue.

  • For ISO Class 5 (formerly Class 100) applications
  • Average surface resistivity of 1012 ohms/cm2
  • Use when handling Class II Category B (≥1000V) static-sensitive devices
  • Measure 2 3/4 in. long (7cm)

Packaged in an electrostatic shield bag. Rolled.