missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cal-Ex™ Decalcifier, Fisher Chemical

Manufacturer:  Fisher Scientific Company CS5101GH

 View more versions of this product

Catalog No. CS510-1D



 

Specifications Description & Specifications

Specifications

100°C
Liquid
2.0
≥98.5 %
MFCD00211743
1L
0.0027 to 0.0038M (1.35 to 1.40N)
7647-01-0,7732-18-5,6381-92-6
Cal-Ex™ Decalcifier
Poly Bottle

Chemical Identifiers

6381-92-6,7647-01-0,7732-18-5
≥98.5 %
MFCD00211743
Cal-Ex™ Decalcifier

Safety and Handling

SDS & Safety Documents

DANGER!

Emergency Overview
Causes burns by all exposure routes. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Do not induce vomiting. Call a physician or Poison Control Center immediately. Call a physician or Poison Control Center immediately. .

NFPA

Health:3
Flammability:0
Instability:0

DOTInformation : DOT Class 8, : Corrosive