missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Caframo™ Stainless Steel Shaft

Passivated for maximum corrosion protection

Manufacturer:  Caframo™ A712

 View more versions of this product

Catalog No. 14-501-072



Description

Description

  • Electro-polished stainless steel 316L

  • 0.375 in. (9.5mm) Shafts have a turned down 0.5 × 0.3125 in. end

Specifications

Specifications

0.2 lb.
0.31 in.
Any propeller with a 0.8cm dia. bore
305 mm
0.09kg
8 mm
12 in.
Shaft
SDS
Documents